Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(2,2,2-Trifluoroethyl)oxirane, 97%, Acros Organics™

Click to view available options
Quantity:
1g
407-12-5
C4H5F3O
126.08
KSAGWVJHDZAMEZ-UHFFFAOYSA-N
2-2,2,2-trifluoroethyl oxirane,2,2,2-trifluoroethyl oxirane,4,4,4-trifluoro-1,2-epoxybutane,acmc-1ctef,1,1,1-trifluoro-3,4-epoxybutane,2-2,2,2-trifluoro-ethyl-oxirane,3-trifluoromethyl-1,2-epoxypropane,oxirane,2-2,2,2-trifluoroethyl,2-2,2,2-tris fluoranyl ethyl oxirane
2782635
2-(2,2,2-trifluoroethyl)oxirane
C1C(O1)CC(F)(F)F
407-12-5
C4H5F3O
126.08
KSAGWVJHDZAMEZ-UHFFFAOYSA-N
2-2,2,2-trifluoroethyl oxirane,2,2,2-trifluoroethyl oxirane,4,4,4-trifluoro-1,2-epoxybutane,acmc-1ctef,1,1,1-trifluoro-3,4-epoxybutane,2-2,2,2-trifluoro-ethyl-oxirane,3-trifluoromethyl-1,2-epoxypropane,oxirane,2-2,2,2-trifluoroethyl,2-2,2,2-tris fluoranyl ethyl oxirane
2782635
2-(2,2,2-trifluoroethyl)oxirane
C1C(O1)CC(F)(F)F
Specific Gravity 1.2487
Density 1.2487g/mL
Boiling Point 68.0°C to 82.0°C
Quantity 1g
Infrared Spectrum Authentic
Flash Point −12°C
Assay Percent Range 96% min. (GC)
Formula Weight 126.08
Percent Purity 97%
Chemical Name or Material (2,2,2-Trifluoroethyl)oxirane

GHS H Statement Highly flammable liquid and vapour. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Danger