Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,2-Trifluoroethyl p-Toluenesulfonate 98.0+%, TCI America™

Click to view available options
Quantity:
25g
500g
433-06-7
C9H9F3O3S
254.223
MFCD00000443
IGKCQDUYZULGBM-UHFFFAOYSA-N
2,2,2-trifluoroethyl p-toluenesulfonate,2,2,2-trifluoroethyl tosylate,trifluoroethanol tosylate,trifluorethyltosylate,2,2,2-trifluoroethyl-p-toluenesulfonate,ethanol, 2,2,2-trifluoro-, 4-methylbenzenesulfonate,2,2,2-trifluoroethyl p-toluenesulphonate,ethanol, 2,2,2-trifluoro-, p-toluenesulfonate,2,2,2-trifluoroethyl 4-methylbenzene-1-sulfonate,ethanol, 2,2,2-trifluoro-, 1-4-methylbenzenesulfonate
67942
2,2,2-trifluoroethyl 4-methylbenzenesulfonate
CC1=CC=C(C=C1)S(=O)(=O)OCC(F)(F)F
433-06-7
C9H9F3O3S
254.223
MFCD00000443
IGKCQDUYZULGBM-UHFFFAOYSA-N
2,2,2-trifluoroethyl p-toluenesulfonate,2,2,2-trifluoroethyl tosylate,trifluoroethanol tosylate,trifluorethyltosylate,2,2,2-trifluoroethyl-p-toluenesulfonate,ethanol, 2,2,2-trifluoro-, 4-methylbenzenesulfonate,2,2,2-trifluoroethyl p-toluenesulphonate,ethanol, 2,2,2-trifluoro-, p-toluenesulfonate,2,2,2-trifluoroethyl 4-methylbenzene-1-sulfonate,ethanol, 2,2,2-trifluoro-, 1-4-methylbenzenesulfonate
67942
2,2,2-trifluoroethyl 4-methylbenzenesulfonate
CC1=CC=C(C=C1)S(=O)(=O)OCC(F)(F)F
Quantity 25g
Boiling Point 104°C
Color White
Melting Point 40°C
Formula Weight 254.22
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (GC,T)
Chemical Name or Material 2,2,2-Trifluoroethyl p-Toluenesulfonate