Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,2-Trifluoroethylhydrazine, 70 wt% aqueous solution, Acros Organics

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
5042-30-8,7732-18-5
C2H5F3N2
114.07
MFCD00007622
OPMFFAOEPFATTG-UHFFFAOYSA-N
2,2,2-trifluoroethyl hydrazine,2,2,2-trifluoroethylhydrazine, 70 % in aqueous solution,2,2,2-trifluoroethyl hydrazine in water,sntljqhjptadtauup@,acmc-20aoks,trifluoroethyl hydrazine,cf3ch2nhnh2,2,2,2-trifluorethylhydrazine
78740
2,2,2-trifluoroethylhydrazine
C(C(F)(F)F)NN
5042-30-8,7732-18-5
C2H5F3N2
114.07
MFCD00007622
OPMFFAOEPFATTG-UHFFFAOYSA-N
2,2,2-trifluoroethyl hydrazine,2,2,2-trifluoroethylhydrazine, 70 % in aqueous solution,2,2,2-trifluoroethyl hydrazine in water,sntljqhjptadtauup@,acmc-20aoks,trifluoroethyl hydrazine,cf3ch2nhnh2,2,2,2-trifluorethylhydrazine
78740
2,2,2-trifluoroethylhydrazine
C(C(F)(F)F)NN
Specific Gravity 1.294
Flash Point 42°C
Refractive Index 1.3600 to 1.364
Color Colorless to Yellow
Quantity 25g
Density 1.2940g/mL
Packaging Glass bottle
CAS Min % 65.0,25.0
CAS Max % 75.0,35.0
Linear Formula CF3CH2NHNH2
Formula Weight 114.07
Physical Form Liquid
Percent Purity ≥65%
Chemical Name or Material 2, 2, 2-Trifluoroethylhydrazine, 70 wt% aqueous solution
Show More Show Less

GHS H Statement
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Flammable liquid and vapor.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Call a POISON CENTER or

GHS Signal Word: Warning

EINECSNumber : 225-734-