Alfa Aesar™ 2,2,2-Trimethoxy-4,5-dimethyl-1,3,2-dioxaphospholene, 97%

Manufacturer: Alfa Aesar™ B2273009

Quantity 10g

 View more versions of this product

Catalog No. AAB2273009

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1665-79-8
    Molecular Formula C7H15O5P
    Formula Weight 210.17
    MDL Number MFCD00014510
    Synonym 2,3-Dihydro-2,2,2-trimethoxy-4,5-dimethyl-1,3,2-dioxaphophole
    Chemical Name or Material 2,2,2-Trimethoxy-4,5-dimethyl-1,3,2-dioxaphospholene

    Specifications

    Formula Weight 210.17
    TSCA TSCA
    Molecular Formula C7H15O5P
    MDL Number MFCD00014510
    Synonym 2,3-Dihydro-2,2,2-trimethoxy-4,5-dimethyl-1,3,2-dioxaphophole
    Boiling Point 64° to 66°C (1mmHg)
    Quantity 10g
    CAS 1665-79-8
    Chemical Name or Material 2,2,2-Trimethoxy-4,5-dimethyl-1,3,2-dioxaphospholene
    Assay 97%

    Description

    Applications
    2,2,2-Trimethoxy-4,5-dimethyl-1,3,2-dioxaphospholene is used as an organic chemical synthesis intermediate.

    Notes
    Stable under recommended storage conditions. Incompatible with oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit