Alfa Aesar™ 2,2,2-Trimethylthioacetamide, 97%

Manufacturer: Alfa Aesar™ H3209209

Quantity 10g

 View more versions of this product

Catalog No. AAH3209209

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 630-22-8
    Molecular Formula C5H11NS
    Formula Weight 117.21
    MDL Number MFCD09742834
    Synonym 2,2-Dimethylpropanethioamide; 2,2-Dimethylthiopropionamide
    Chemical Name or Material 2,2,2-Trimethylthioacetamide

    Specifications

    Formula Weight 117.21
    MDL Number MFCD09742834
    CAS 630-22-8
    Synonym 2,2-Dimethylpropanethioamide; 2,2-Dimethylthiopropionamide
    Health Hazard 2 GHS H Statement
    H301
    Toxic if swallowed.
    Quantity 10g
    Molecular Formula C5H11NS
    Health Hazard 1 Danger
    Health Hazard 3 GHS P Statement
    P264-P270-P301+P310-P321-P405-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Melting Point 113° to 117°C
    Chemical Name or Material 2,2,2-Trimethylthioacetamide
    Assay 97%

    Description

    Applications
    2,2,2-Trimethylthioacetamide, is used as a pharmaceutical and organic intermediate, an important amino protective agent, pharmaceutical and synthetic organic make intermediates, organic synthesis used to introduce t- Boc protect gene species.

    Solubility
    Soluble in water (10 g/L) (25°C).

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

    GHS H Statement
    H301
    Toxic if swallowed.

    GHS P Statement
    P264-P270-P301+P310-P321-P405-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit