Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,2-Triphenylacetophenone 99.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
466-37-5
C26H20O
348.445
MFCD00004762
CFBBKHROQRFCNZ-UHFFFAOYSA-N
2,2,2-triphenylacetophenone,tetraphenylethanone,benzopinacolone,ethanone, tetraphenyl,phenyl trityl ketone,acetophenone, 2,2,2-triphenyl,1,2,2,2-tetraphenylethan-1-one,ss-benzpinakolin,beta-benzopinacolone,.beta.-benzopinacolone
68049
1,2,2,2-tetraphenylethanone
C1=CC=C(C=C1)C(=O)C(C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4
466-37-5
C26H20O
348.445
MFCD00004762
CFBBKHROQRFCNZ-UHFFFAOYSA-N
2,2,2-triphenylacetophenone,tetraphenylethanone,benzopinacolone,ethanone, tetraphenyl,phenyl trityl ketone,acetophenone, 2,2,2-triphenyl,1,2,2,2-tetraphenylethan-1-one,ss-benzpinakolin,beta-benzopinacolone,.beta.-benzopinacolone
68049
1,2,2,2-tetraphenylethanone
C1=CC=C(C=C1)C(=O)C(C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4
Quantity 25g
Color White
Melting Point 184°C
Formula Weight 348.45
Physical Form Crystal-Powder at 20°C
Percent Purity ≥99.0% (GC)
Chemical Name or Material 2,2,2-Triphenylacetophenone