Alfa Aesar™ 2,2,3,3,3-Pentafluoro-1-propanol, 96%

Manufacturer: Alfa Aesar™ 1587309

Quantity 10g

 View more versions of this product

Catalog No. AA1587309

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 422-05-9
    Formula Weight 150.05
    MDL Number MFCD00004673
    Beilstein 1743133
    Synonym 1H,1H-Pentafluoro-1-propanol
    Chemical Name or Material 2,2,3,3,3-Pentafluoro-1-propanol

    Specifications

    Formula Weight 150.05
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Boiling Point 80° to 81°C
    RTECS Number UB8800000
    Density 1.505g/mL
    Refractive Index 1.2880
    Physical Form Liquid
    Quantity 10g
    MDL Number MFCD00004673
    Synonym 1H,1H-Pentafluoro-1-propanol
    Health Hazard 1 Warning
    TSCA TSCA
    CAS 422-05-9
    Beilstein 1743133
    Health Hazard 2 GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Chemical Name or Material 2,2,3,3,3-Pentafluoro-1-propanol
    Assay 96%

    GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit