Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,2,3,3,3-Pentafluoropropyl methacrylate, 97%, stab.

Click to view available options
Quantity:
5g
25g
45115-53-5
C7H7F5O2
218.123
MFCD00039256
CLISWDZSTWQFNX-UHFFFAOYSA-N
1h,1h-pentafluoropropyl methacrylate,2,2,3,3,3-pentafluoropropyl methacrylate,2-propenoic acid, 2-methyl-, 2,2,3,3,3-pentafluoropropyl ester,1h,1h-pentafluoro-n-propyl methacrylate,2,2,3,3,3-pentafluoro-n-propyl methacrylate,methacrylic acid 2,2,3,3,3-pentafluoropropyl ester,2,2,3,3,3-pentafluoropropyl methacrylate stabilized with tbc,2,2,3,3,3-pentafluoropropyl methacrylate, contains 100 ppm 4-tert-butylcatechol as inhibitor
123516
2,2,3,3,3-pentafluoropropyl 2-methylprop-2-enoate
CC(=C)C(=O)OCC(C(F)(F)F)(F)F
45115-53-5
C7H7F5O2
218.123
MFCD00039256
CLISWDZSTWQFNX-UHFFFAOYSA-N
1h,1h-pentafluoropropyl methacrylate,2,2,3,3,3-pentafluoropropyl methacrylate,2-propenoic acid, 2-methyl-, 2,2,3,3,3-pentafluoropropyl ester,1h,1h-pentafluoro-n-propyl methacrylate,2,2,3,3,3-pentafluoro-n-propyl methacrylate,methacrylic acid 2,2,3,3,3-pentafluoropropyl ester,2,2,3,3,3-pentafluoropropyl methacrylate stabilized with tbc,2,2,3,3,3-pentafluoropropyl methacrylate, contains 100 ppm 4-tert-butylcatechol as inhibitor
123516
2,2,3,3,3-pentafluoropropyl 2-methylprop-2-enoate
CC(=C)C(=O)OCC(C(F)(F)F)(F)F
Flash Point 29°C (84°F)
Quantity 5g
Boiling Point 54°C to 55°C (100mmHg)
UN Number UN3272
Formula Weight 218.12
Percent Purity 97%
Chemical Name or Material 2,2,3,3,3-Pentafluoropropyl methacrylate, stabilized

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.

TSCA : TSCA

Recommended Storage : 2° to 8°C