Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,3,3,4,4,4-Heptafluorobutylamine, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
374-99-2
C4H4F7N
199.07
MFCD00014817
WBGBQSRNXPVFDB-UHFFFAOYSA-N
2,2,3,3,4,4,4-heptafluorobutylamine,1h,1h-heptafluorobutylamine,heptafluorobutylamine,butylamine, 2,2,3,3,4,4,4-heptafluoro,1-butanamine, 2,2,3,3,4,4,4-heptafluoro,1h,1h-perfluorobutylamine,acmc-1af94,1-butanamine,2,2,3,3,4,4,4-heptafluoro
67807
2,2,3,3,4,4,4-heptafluorobutan-1-amine
C(C(C(C(F)(F)F)(F)F)(F)F)N
374-99-2
C4H4F7N
199.07
MFCD00014817
WBGBQSRNXPVFDB-UHFFFAOYSA-N
2,2,3,3,4,4,4-heptafluorobutylamine,1h,1h-heptafluorobutylamine,heptafluorobutylamine,butylamine, 2,2,3,3,4,4,4-heptafluoro,1-butanamine, 2,2,3,3,4,4,4-heptafluoro,1h,1h-perfluorobutylamine,acmc-1af94,1-butanamine,2,2,3,3,4,4,4-heptafluoro
67807
2,2,3,3,4,4,4-heptafluorobutan-1-amine
C(C(C(C(F)(F)F)(F)F)(F)F)N
Boiling Point 70°C to 71°C
Specific Gravity 1.493
Density 1.4930g/mL
Refractive Index 1.2960 to 1.298
Quantity 1g
Color Colorless to Yellow
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Linear Formula CF3CF2CF2CH2NH2
Formula Weight 199.07
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 2,2,3,3,4,4,4-Heptafluorobutylamine, 97%
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 206-780-