Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2,2,3,3,4,4-Hexafluoropentanedioic Anhydride 97.0+%, TCI America™
Click to view available options
Quantity:
1g
5g
25g
Specifications
Quantity | 25g |
Color | Yellow |
Boiling Point | 72°C |
Formula Weight | 222.04 |
Physical Form | Clear Liquid at 20°C |
Percent Purity | ≥97.0% (T) |
Chemical Name or Material | 2,2,3,3,4,4-Hexafluoropentanedioic Anhydride |
Safety and Handling
RTECSNumber : TM0860000
TSCA : No