Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,3,3-Tetrafluoro-1-propanol, 97%, Thermo Scientific Chemicals

Catalog Number aab2159409
Click to view available options
Quantity:
10 g
50 g
250 g
76-37-9
C3H4F4O
132.06
MFCD00004676
NBUKAOOFKZFCGD-UHFFFAOYSA-N
2,2,3,3-tetrafluoro-1-propanol, 2,2,3,3-tetrafluoropropanol, 1h,1h,3h-tetrafluoro-1-propanol, 1-propanol, 2,2,3,3-tetrafluoro, tetrafluoropropyl alcohol, 2,2,3,3-tetrafluoropropyl alcohol, 1,1,3-trihydroperfluoro-1-propanol, unii-rno513716p, 1h,1h,3h-tetrafluoropropanol, wln: q1xffyff
6441
2,2,3,3-tetrafluoropropan-1-ol
OCC(F)(F)C(F)F
76-37-9
C3H4F4O
132.06
MFCD00004676
NBUKAOOFKZFCGD-UHFFFAOYSA-N
2,2,3,3-tetrafluoro-1-propanol, 2,2,3,3-tetrafluoropropanol, 1h,1h,3h-tetrafluoro-1-propanol, 1-propanol, 2,2,3,3-tetrafluoro, tetrafluoropropyl alcohol, 2,2,3,3-tetrafluoropropyl alcohol, 1,1,3-trihydroperfluoro-1-propanol, unii-rno513716p, 1h,1h,3h-tetrafluoropropanol, wln: q1xffyff
6441
2,2,3,3-tetrafluoropropan-1-ol
OCC(F)(F)C(F)F

2,2,3,3-Tetrafluoro-1-propanol, is used as an essential fluorinated fine chemical, the main application of TFP is as a dye solvent in CD-R(W) and DVD-R(W) industry. It is also used in making textile treatment agent and pharmaceuticals. This chemical is an ideal replacement for Freon 22 detergent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2,3,3-Tetrafluoro-1-propanol, is used as an essential fluorinated fine chemical, the main application of TFP is as a dye solvent in CD-R(W) and DVD-R(W) industry. It is also used in making textile treatment agent and pharmaceuticals. This chemical is an ideal replacement for Freon 22 detergent.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point <-70°C
Density 1.471
Boiling Point 107°C to 109°C
Flash Point 49°C (120°F)
Refractive Index 1.3195
Quantity 10 g
UN Number UN1987
Beilstein 1738264
Merck Index 14,9206
Solubility Information Soluble in water.
Formula Weight 132.06
Percent Purity 97%
Chemical Name or Material 2,2,3,3-Tetrafluoro-1-propanol
Show More Show Less
Hazard Category H226-H315-H319-H332-H335
Hazard Statement GHS H Statement
H226-H332-H315-H319-H335
Flammable liquid and vapour.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
EINECSNumber 200-955-5
RTECSNumber UB9400000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit