Alfa Aesar™ 2,2,3,3-Tetrafluoropropyl methacrylate, 97%, stab. with 50ppm 4-methoxyphenol

Manufacturer: Alfa Aesar™ B22682-14

Quantity 25g

 View more versions of this product

Catalog No. AAB2268214

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 45102-52-1
    Molecular Formula C7H8F4O2
    Formula Weight 200.13g/mol
    MDL Number MFCD00042381
    UN Number UN3272
    Beilstein 3588831
    Synonym Methacrylic acid 2,2,3,3-tetrafluoropropyl ester; 2,2,3,3-Tetrafluoropropyl 2-methylpropenoate
    Chemical Name or Material 2,2,3,3-Tetrafluoropropyl methacrylate, stabilized with 50ppm 4-methoxyphenol

    Specifications

    Boiling Point 146° to 149°C
    Flash Point 50°C (122°F)
    Quantity 25g
    CAS 45102-52-1
    Molecular Formula C7H8F4O2
    Formula Weight 200.13g/mol
    MDL Number MFCD00042381
    UN Number UN3272
    Beilstein 3588831
    Synonym Methacrylic acid 2,2,3,3-tetrafluoropropyl ester; 2,2,3,3-Tetrafluoropropyl 2-methylpropenoate
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H226-H315-H319-H335
    Flammable liquid and vapour.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
    Chemical Name or Material 2,2,3,3-Tetrafluoropropyl methacrylate, stabilized with 50ppm 4-methoxyphenol
    Assay 97%

    GHS H Statement
    H226-H315-H319-H335
    Flammable liquid and vapour.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit