Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,2,3,3-Tetrafluoropropyl methacrylate, 97%, stab. with 50ppm 4-methoxyphenol

Click to view available options
Quantity:
5g
25g
45102-52-1
C7H8F4O2
200.133
MFCD00042381
RSVZYSKAPMBSMY-UHFFFAOYSA-N
2,2,3,3-tetrafluoropropyl methacrylate,2-propenoic acid, 2-methyl-, 2,2,3,3-tetrafluoropropyl ester,methacrylic acid 2,2,3,3-tetrafluoropropyl ester,1h,1h,3h-tetrafluoropropyl methacrylate,2,2,3,3-tetrafluoropropyl methacrylate, stab. with 2,2'-methylenebis 6-tert-butyl-4-methyl-phenol,acmc-1ajz7,ksc489s8d,rsvzyskapmbsmy-uhfffaoysa,2,2,3,3-tetrafluoroprop-1-yl methacrylat
123515
2,2,3,3-tetrafluoropropyl 2-methylprop-2-enoate
CC(=C)C(=O)OCC(C(F)F)(F)F
45102-52-1
C7H8F4O2
200.133
MFCD00042381
RSVZYSKAPMBSMY-UHFFFAOYSA-N
2,2,3,3-tetrafluoropropyl methacrylate,2-propenoic acid, 2-methyl-, 2,2,3,3-tetrafluoropropyl ester,methacrylic acid 2,2,3,3-tetrafluoropropyl ester,1h,1h,3h-tetrafluoropropyl methacrylate,2,2,3,3-tetrafluoropropyl methacrylate, stab. with 2,2'-methylenebis 6-tert-butyl-4-methyl-phenol,acmc-1ajz7,ksc489s8d,rsvzyskapmbsmy-uhfffaoysa,2,2,3,3-tetrafluoroprop-1-yl methacrylat
123515
2,2,3,3-tetrafluoropropyl 2-methylprop-2-enoate
CC(=C)C(=O)OCC(C(F)F)(F)F
Quantity 5g
Flash Point 50°C (122°F)
Boiling Point 146°C to 149°C
UN Number UN3272
Beilstein 3588831
Formula Weight 200.13
Percent Purity 97%
Chemical Name or Material 2,2,3,3-Tetrafluoropropyl methacrylate, stabilized with 50ppm 4-methoxyphenol

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.