Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,3,3-Tetramethylbutane 98.0+%, TCI America™

Click to view available options
1G
594-82-1
C8H18
114.232
MFCD00008849
OMMLUKLXGSRPHK-UHFFFAOYSA-N
Hexamethylethane
11675
2,2,3,3-tetramethylbutane
CC(C)(C)C(C)(C)C
594-82-1
C8H18
114.232
MFCD00008849
OMMLUKLXGSRPHK-UHFFFAOYSA-N
Hexamethylethane
11675
2,2,3,3-tetramethylbutane
CC(C)(C)C(C)(C)C
Melting Point 100°C
Color White
Quantity 1g
UN Number 1325
Formula Weight 114.23
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (GC)
Chemical Name or Material 2,2,3,3-Tetramethylbutane

EINECSNumber : (2)-0008

TSCA : No