Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,3,4,4,4-Hexafluorobutyl Acrylate (stabilized with TBC) 98.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
54052-90-3
C7H6F6O2
236.113
MFCD00044091
LMVLEDTVXAGBJV-UHFFFAOYSA-N
Acrylic Acid 2,2,3,4,4,4-Hexafluorobutyl Ester, HFBA
2774977
2,2,3,4,4,4-hexafluorobutyl prop-2-enoate
C=CC(=O)OCC(C(C(F)(F)F)F)(F)F
54052-90-3
C7H6F6O2
236.113
MFCD00044091
LMVLEDTVXAGBJV-UHFFFAOYSA-N
Acrylic Acid 2,2,3,4,4,4-Hexafluorobutyl Ester, HFBA
2774977
2,2,3,4,4,4-hexafluorobutyl prop-2-enoate
C=CC(=O)OCC(C(C(F)(F)F)F)(F)F
Quantity 25g
Color Colorless
Boiling Point 43°C
UN Number 3272
Formula Weight 236.11
Physical Form Clear Liquid at 20°C
Percent Purity ≥98.0% (GC)
Chemical Name or Material 2,2,3,4,4,4-Hexafluorobutyl Acrylate (stabilized with TBC)

TSCA : No

Recommended Storage : Refrigerator