Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(2,3-Dihydro-1-benzofuran-5-yl)-1,3-thiazole-4-carboxylic acid, 97%, Thermo Scientific™

Catalog Number SP00242DA
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
368869-97-0
C12H9NO3S
247.27
MFCD01313485
NOUOFPSRFBTJCH-UHFFFAOYSA-N
2-2,3-dihydro-1-benzofuran-5-yl-1,3-thiazole-4-carboxylic acid, 2-2,3-dihydrobenzofuran-5-yl thiazole-4-carboxylic acid, 4-thiazolecarboxylicacid, 2-2,3-dihydro-5-benzofuranyl, maybridge1_008369, 2-2,3-dihydrobenzofuran-5-yl-4-thiazolecarboxylic acid, 2-2,3-dihydrobenzo b furan-5-yl-1,3-thiazole-4-carboxylic acid, 2-2,3-dihydro-1-benzo b furan-5-yl-1,3-thiazole-4-carboxylic acid
2742272
2-(2,3-dihydro-1-benzofuran-5-yl)-1,3-thiazole-4-carboxylic acid
OC(=O)C1=CSC(=N1)C1=CC2=C(OCC2)C=C1
368869-97-0
C12H9NO3S
247.27
MFCD01313485
NOUOFPSRFBTJCH-UHFFFAOYSA-N
2-2,3-dihydro-1-benzofuran-5-yl-1,3-thiazole-4-carboxylic acid, 2-2,3-dihydrobenzofuran-5-yl thiazole-4-carboxylic acid, 4-thiazolecarboxylicacid, 2-2,3-dihydro-5-benzofuranyl, maybridge1_008369, 2-2,3-dihydrobenzofuran-5-yl-4-thiazolecarboxylic acid, 2-2,3-dihydrobenzo b furan-5-yl-1,3-thiazole-4-carboxylic acid, 2-2,3-dihydro-1-benzo b furan-5-yl-1,3-thiazole-4-carboxylic acid
2742272
2-(2,3-dihydro-1-benzofuran-5-yl)-1,3-thiazole-4-carboxylic acid
OC(=O)C1=CSC(=N1)C1=CC2=C(OCC2)C=C1
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 247.27
Percent Purity 97%
Chemical Name or Material 2-(2, 3-Dihydro-1-benzofuran-5-yl)-1, 3-thiazole-4-carboxylic acid
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit