Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,4,4,6,6,8,8-Octamethylcyclotetrasilazane, 95%, ACROS Organics™

Click to view available options
Quantity:
25g
1020-84-4
C8H28N4Si4
MFCD00046123
1020-84-4
C8H28N4Si4
MFCD00046123
Quantity 25g
Melting Point 94°C to 99°C
Color White
Infrared Spectrum Authentic
Density 0.95g/mL
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Formula Weight 292.68
Physical Form Crystals
Percent Purity ≥94%
Chemical Name or Material 2,2,4,4,6,6,8,8-Octamethylcyclotetrasilazane, 95%
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Toxic if swallowed.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 213-817-4

RTECSNumber : GZ4342500

TSCA : TSCA