Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,2,4,4-Tetrafluoro-1,3-dithietane, 97%

Click to view available options
Quantity:
10g
50g
1717-50-6
C2F4S2
164.136
MFCD00053538
MPOCEMBRPHOCMK-UHFFFAOYSA-N
tetrafluoro-1,3-dithietane,pubchem3989,acmc-1budt,1, 2,2,4,4-tetrafluoro,2,4,4-tetrafluoro-1,3-dithietane,1,3-dithietane,2,2,4,4-tetrafluoro,2,2,4,4-tetrafluoro-1,3-dithietane
74392
2,2,4,4-tetrafluoro-1,3-dithietane
C1(SC(S1)(F)F)(F)F
1717-50-6
C2F4S2
164.136
MFCD00053538
MPOCEMBRPHOCMK-UHFFFAOYSA-N
tetrafluoro-1,3-dithietane,pubchem3989,acmc-1budt,1, 2,2,4,4-tetrafluoro,2,4,4-tetrafluoro-1,3-dithietane,1,3-dithietane,2,2,4,4-tetrafluoro,2,2,4,4-tetrafluoro-1,3-dithietane
74392
2,2,4,4-tetrafluoro-1,3-dithietane
C1(SC(S1)(F)F)(F)F
Quantity 10g
Boiling Point 47°C to 49°C
Flash Point 36°C (96°F)
Odor Unpleasant
UN Number UN1993
Formula Weight 164.14
Percent Purity 97%
Chemical Name or Material 2,2,4,4-Tetrafluoro-1,3-dithietane

GHS H Statement
H226
Flammable liquid and vapour.

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.