Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,4,4-Tetramethyl-3-pentanone, 98%, Thermo Scientific™

Catalog No. AC434760010
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle

Chemical Identifiers

CAS 815-24-7
Molecular Formula C9H18O
Molecular Weight (g/mol) 142.24
MDL Number MFCD00008845
InChI Key UIQGEWJEWJMQSL-UHFFFAOYSA-N
Synonym hexamethylacetone, 2,2,4,4-tetramethyl-3-pentanone, di-tert-butyl ketone, 3-pentanone, 2,2,4,4-tetramethyl, 2,2,4,4-tetramethyl-pentan-3-one, 3-pentanone,2,2,4,4-tetramethyl, di-t-butyl ketone, acmc-20akrk, t-c4h9 2co, 2,4,4-tetramethyl-3-pentanone
PubChem CID 13152
IUPAC Name 2,2,4,4-tetramethylpentan-3-one
SMILES CC(C)(C)C(=O)C(C)(C)C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 0.8240g/mL
Boiling Point 150.0°C to 155.0°C
Flash Point 34°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Specific Gravity 0.824
Formula Weight 142.24
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2,2,4,4-Tetramethyl-3-pentanone
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Flammable liquid and vapour.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
EINECSNumber 212-419-8

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.