Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,4,5,5-Pentamethyl-3-imidazoline-3-oxide-1-oxyl, Free Radical 98%, ACROS Organics™

Click to view available options
Quantity:
250mg
Packaging:
Glass bottle
18796-04-8
C8 H15 N2 O2
MFCD00078169
18796-04-8
C8 H15 N2 O2
MFCD00078169
Polymerization reagent
Color Orange to Brown
Infrared Spectrum Authentic
Quantity 250mg
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 171.22
Physical Form Crystalline Powder or Chunks
Percent Purity ≥96.0%
Chemical Name or Material 2,2,4,5,5-Pentamethyl-3-imidazoline-3-oxide-1-oxyl, 98%
Show More Show Less