Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,4,5,5-Pentamethyl-3-imidazoline-3-oxide, Free Radical 98%, ACROS Organics™

Click to view available options
Quantity:
100mg
Packaging:
Glass bottle
64934-83-4
C8H16N2O
156.229
MFCD00078168
GMIPCEOKROIPSM-UHFFFAOYSA-N
2,2,4,5,5-pentamethyl-3-imidazoline-3-oxide,ambscl14/014,2,2,4,5,5-pentamethyl-3-imidazoline 3-oxide,2,2,4,4,5-pentamethyl-3h-imidazol-1-ium-1-olate,1h-imidazole,2,5-dihydro-2,2,4,5,5-pentamethyl-, 3-oxide
547136
2,2,4,5,5-pentamethyl-3-oxido-1H-imidazol-3-ium
CC1=[N+](C(NC1(C)C)(C)C)[O-]
64934-83-4
C8H16N2O
156.229
MFCD00078168
GMIPCEOKROIPSM-UHFFFAOYSA-N
2,2,4,5,5-pentamethyl-3-imidazoline-3-oxide,ambscl14/014,2,2,4,5,5-pentamethyl-3-imidazoline 3-oxide,2,2,4,4,5-pentamethyl-3h-imidazol-1-ium-1-olate,1h-imidazole,2,5-dihydro-2,2,4,5,5-pentamethyl-, 3-oxide
547136
2,2,4,5,5-pentamethyl-3-oxido-1H-imidazol-3-ium
CC1=[N+](C(NC1(C)C)(C)C)[O-]
Packaging Glass bottle
Quantity 100mg
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 156.23
Chemical Name or Material 2,2,4,5,5-Pentamethyl-3-imidazoline-3-oxide, 98%