Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(2,4,5-Trichlorophenoxy)propionic acid, 96%, Thermo Scientific™

Click to view available options
Quantity:
5g
25g
93-72-1
C9H7Cl3O3
269.50
MFCD00002646
ZLSWBLPERHFHIS-UHFFFAOYNA-N
fenoprop, silvex, 2-2,4,5-trichlorophenoxy propionic acid, double strength, fenormone, propon, color-set, aqua-vex, sta-fast, fruitone t
7158
CHEBI:34758
2-(2,4,5-trichlorophenoxy)propanoic acid
CC(OC1=CC(Cl)=C(Cl)C=C1Cl)C(O)=O
93-72-1
C9H7Cl3O3
269.50
MFCD00002646
ZLSWBLPERHFHIS-UHFFFAOYNA-N
fenoprop, silvex, 2-2,4,5-trichlorophenoxy propionic acid, double strength, fenormone, propon, color-set, aqua-vex, sta-fast, fruitone t
7158
CHEBI:34758
2-(2,4,5-trichlorophenoxy)propanoic acid
CC(OC1=CC(Cl)=C(Cl)C=C1Cl)C(O)=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Melting Point 176°C to 180°C
UN Number UN3077
Beilstein 1985768
Quantity 25g
Formula Weight 269.51
Percent Purity 96%
Chemical Name or Material 2-(2,4,5-Trichlorophenoxy)propionic acid

GHS H Statement
H302-H315
Harmful if swallowed.
Causes skin irritation.

P201-P202-P264b-P270-P280i-P281-P301+P312-P302+P352-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c

H302-H315-H319-H351

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

EINECSNumber : 202-271-2

RTECSNumber : UF8225000

TSCA : No

Recommended Storage : Ambient temperatures