Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(2,4,5-Trichlorophenoxy)propionic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
93-72-1
C9H7Cl3O3
269.502
MFCD00002646
ZLSWBLPERHFHIS-UHFFFAOYSA-N
fenoprop,silvex,2-2,4,5-trichlorophenoxy propionic acid,double strength,fenormone,propon,color-set,aqua-vex,sta-fast,fruitone t
7158
CHEBI:34758
2-(2,4,5-trichlorophenoxy)propanoic acid
CC(C(=O)O)OC1=CC(=C(C=C1Cl)Cl)Cl
93-72-1
C9H7Cl3O3
269.502
MFCD00002646
ZLSWBLPERHFHIS-UHFFFAOYSA-N
fenoprop,silvex,2-2,4,5-trichlorophenoxy propionic acid,double strength,fenormone,propon,color-set,aqua-vex,sta-fast,fruitone t
7158
CHEBI:34758
2-(2,4,5-trichlorophenoxy)propanoic acid
CC(C(=O)O)OC1=CC(=C(C=C1Cl)Cl)Cl
Melting Point 177°C to 181°C
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 5g
Color Beige
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula CH3CH(OC6H2Cl3)CO2H
Merck Index 13, 8606
Formula Weight 269.51
Physical Form Fine Crystalline Powder
Percent Purity ≥96.0%
Chemical Name or Material 2-(2, 4, 5-Trichlorophenoxy)propionic acid, 97%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
Very toxic to aquatic life with long lasting effects.

GHS P Statement:
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning