Alfa Aesar™ 2-(2,4-Dichlorophenoxy)propionic acid, 98%

Manufacturer: Alfa Aesar™ B2434422

Quantity 100g

 View more versions of this product

Catalog No. AAB2434422

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 120-36-5
    Molecular Formula C9H8Cl2O3
    Formula Weight 235.07
    MDL Number MFCD00002645
    Beilstein 2213812
    Merck Index 14,3078
    Synonym Dichlorprop; 2,4-DP
    Chemical Name or Material 2-(2,4-Dichlorophenoxy)propionic acid

    Specifications

    Formula Weight 235.07
    TSCA TSCA
    Odor Odorless
    Molecular Formula C9H8Cl2O3
    MDL Number MFCD00002645
    Beilstein 2213812
    Melting Point 112° to 116°C
    Merck Index 14,3078
    Synonym Dichlorprop; 2,4-DP
    Health Hazard 2 GHS H Statement
    H318-H302-H312-H315
    Causes serious eye damage.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    RTECS Number UF1050000
    Quantity 100g
    CAS 120-36-5
    Health Hazard 1 Danger
    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P310-P362-P301+P312-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    Take off contaminated clothing and wash before reuse.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material 2-(2,4-Dichlorophenoxy)propionic acid
    Assay 98%

    GHS H Statement
    H318-H302-H312-H315
    Causes serious eye damage.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.

    GHS P Statement
    P280-P305+P351+P338-P310-P362-P301+P312-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    Take off contaminated clothing and wash before reuse.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit