Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-(2,4-Dichlorophenoxy)propionic acid, 98%

Click to view available options
Quantity:
100g
500g
120-36-5
C9H8Cl2O3
235.06
MFCD00002645
MZHCENGPTKEIGP-UHFFFAOYSA-N
dichlorprop,2-2,4-dichlorophenoxy propanoic acid,2-2,4-dichlorophenoxy propionic acid,hormatox,polymone,dichloroprop,polytox,kildip,celatox-dp,herbizid dp
8427
CHEBI:75370
2-(2,4-dichlorophenoxy)propanoic acid
CC(C(=O)O)OC1=C(C=C(C=C1)Cl)Cl
120-36-5
C9H8Cl2O3
235.06
MFCD00002645
MZHCENGPTKEIGP-UHFFFAOYSA-N
dichlorprop,2-2,4-dichlorophenoxy propanoic acid,2-2,4-dichlorophenoxy propionic acid,hormatox,polymone,dichloroprop,polytox,kildip,celatox-dp,herbizid dp
8427
CHEBI:75370
2-(2,4-dichlorophenoxy)propanoic acid
CC(C(=O)O)OC1=C(C=C(C=C1)Cl)Cl
Quantity 500g
Odor Odorless
Melting Point 112°C to 116°C
Beilstein 2213812
Merck Index 14,3078
Formula Weight 235.07
Percent Purity 98%
Chemical Name or Material 2-(2,4-Dichlorophenoxy)propionic acid

GHS H Statement
H318-H302-H312-H315
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.

GHS P Statement
P280-P305+P351+P338-P310-P362-P301+P312-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Take off contaminated clothing and wash before reuse.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : UF1050000

TSCA : TSCA