Alfa Aesar™ 2,2',4'-Trichloroacetophenone, 97%

Manufacturer: Alfa Aesar™ A1083514

Quantity 25g

 View more versions of this product

Catalog No. AAA1083514

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 4252-78-2
    Molecular Formula C8H5Cl3O
    Formula Weight 223.49
    MDL Number MFCD00000934
    UN Number UN2923
    Beilstein 957098
    Synonym 2,4-Dichlorophenacyl chloride; alpha,2,4-Trichloroacetophenone
    Chemical Name or Material 2,2',4'-Trichloroacetophenone

    Specifications

    Formula Weight 223.49
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 4252-78-2
    Health Hazard 1 Danger
    Quantity 25g
    Molecular Formula C8H5Cl3O
    Beilstein 957098
    Synonym 2,4-Dichlorophenacyl chloride; alpha,2,4-Trichloroacetophenone
    Health Hazard 2 GHS H Statement
    H301-H314-H318-H335
    Toxic if swallowed.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    May cause respiratory irritation.
    Melting Point 52° to 56°C
    Flash Point >110°C (230°F)
    MDL Number MFCD00000934
    UN Number UN2923
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
    Chemical Name or Material 2,2',4'-Trichloroacetophenone
    Assay 97%

    GHS H Statement
    H301-H314-H318-H335
    Toxic if swallowed.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    May cause respiratory irritation.

    GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit