Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,2,4-Trimethyl-1,3-dioxolane, 99%

Click to view available options
Quantity:
25g
100g
500g
1193-11-9
C6H12O2
116.16
MFCD00090841
ALTFLAPROMVXNX-UHFFFAOYSA-N
propylene glycol acetone ketal,1,3-dioxolane, 2,2,4-trimethyl,acetone propylene glycol acetal,1,3-dioxolane,2,2,4-trimethyl,fema no. 3441,2,2,4-trimethyl-1,3-oxacyclopentane,2,2,4-trimethyl-1,3-dioxacyclopentane,propylene glycol acetonide,1,2-propanediol acetonide,2,2,4-trimethyldioxolane
62384
2,2,4-trimethyl-1,3-dioxolane
CC1COC(O1)(C)C
1193-11-9
C6H12O2
116.16
MFCD00090841
ALTFLAPROMVXNX-UHFFFAOYSA-N
propylene glycol acetone ketal,1,3-dioxolane, 2,2,4-trimethyl,acetone propylene glycol acetal,1,3-dioxolane,2,2,4-trimethyl,fema no. 3441,2,2,4-trimethyl-1,3-oxacyclopentane,2,2,4-trimethyl-1,3-dioxacyclopentane,propylene glycol acetonide,1,2-propanediol acetonide,2,2,4-trimethyldioxolane
62384
2,2,4-trimethyl-1,3-dioxolane
CC1COC(O1)(C)C
Boiling Point 98°C
Quantity 25g
Flash Point 32°C (90°F)
UN Number UN3271
Formula Weight 116.16
Percent Purity 99%
Chemical Name or Material 2,2,4-Trimethyl-1,3-dioxolane

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHERS, N.O.S.