Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 2,2,4-Trimethyl-1,3-dioxolane, 99%
Click to view available options
Quantity:
25g
100g
500g
Specifications
Flash Point | 32°C (90°F) |
Quantity | 100g |
Density | 0.897 |
Boiling Point | 98°C |
UN Number | UN3271 |
Formula Weight | 116.16 |
Percent Purity | 99% |
Chemical Name or Material | 2,2,4-Trimethyl-1,3-dioxolane |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHERS, N.O.S.
EINECSNumber : 214-766-0
TSCA : No
Recommended Storage : Ambient temperatures