Learn More
2,2,4-Trimethylpentane, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™, ACROS Organics™
Specifications
CAS Min % | 99.4 |
CAS Max % | 100.0 |
Acetic Acid | 0.001% max. |
Assay Percent Range | 99.5% |
Linear Formula | (CH3)2CHCH2C(CH3)3 |
Beilstein | 01, IV, 439 |
Merck Index | 15, 5239 |
Formula Weight | 114.23 |
Physical Form | Liquid |
Percent Purity | 99.5% |
Show More |
Safety and Handling
GHS H Statement:
May be fatal if swallowed and enters airways.
Very toxic to aquatic life with long lasting effects.
May cause drowsiness or dizziness.
Causes skin irritation.
Highly flammable liquid and vapor.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 208-759-1
RTECSNumber : SA3320000
TSCA : TSCA