Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,4-Trimethylpentane, 99%, for spectroscopy, Thermo Scientific Chemicals

Catalog Number p-216662
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Glass Bottle
540-84-1
C8H18
114.232
MFCD00008943
NHTMVDHEPJAVLT-UHFFFAOYSA-N
isooctane, isobutyltrimethylmethane, pentane, 2,2,4-trimethyl, iso-octane, 2,4,4-trimethylpentane, unii-qab8f5669o, ch3 2chch2c ch3 3, 2, 2, 4-trimethylpentane, i-octane, isobutyltrimethylethane
10907
CHEBI:62805
2,2,4-trimethylpentane
CC(C)CC(C)(C)C
540-84-1
C8H18
114.232
MFCD00008943
NHTMVDHEPJAVLT-UHFFFAOYSA-N
isooctane, isobutyltrimethylmethane, pentane, 2,2,4-trimethyl, iso-octane, 2,4,4-trimethylpentane, unii-qab8f5669o, ch3 2chch2c ch3 3, 2, 2, 4-trimethylpentane, i-octane, isobutyltrimethylethane
10907
CHEBI:62805
2,2,4-trimethylpentane
CC(C)CC(C)(C)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For UV/VIS spectroscopy
TRUSTED_SUSTAINABILITY
Melting Point -107.0°C
Absorbance 0.01 max. at 250nm, 0.01 max. at 400nm, 0.04 max. at 240nm, 0.1 max. at 230nm, 0.2 max. at 220nm, 1 max. at 210nm
CAS Min % 98.5
CAS Max % 100.0
Density 0.6900g/mL
Boiling Point 98.0°C to 99.0°C
Quantity 500 mL
Flash Point −12°C
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.3915
Residue after Evaporation 0.001% max.
Assay Percent Range 99%
Specific Gravity 0.69
Linear Formula (CH3)2CHCH2C(CH3)3
Viscosity 0.51 mPa.s (22°C)
Beilstein 01, IV, 439
Water 0.02% max.
Merck Index 15, 5239
Solubility Information Solubility in water: insoluble. Other solubilities: slightly soluble in alcohol, soluble in benzene, toluene, xylene, chloroform,, dmf, ether, ccl4 and cs2, miscible with acetone, soluble in oils except castor oil
Formula Weight 114.23
Percent Purity 99%
Grade Spectroscopy
Chemical Name or Material 2, 2, 4-Trimethylpentane, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
May be fatal if swallowed and enters airways.
Very toxic to aquatic life with long lasting effects.
May cause drowsiness or dizziness.
Causes skin irritation.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 208-759-1
RTECSNumber SA3320000
TSCA TSCA

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit