Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,4-Trimethylpentane, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500mL
4L
Packaging:
Narrow Mouth Amber Glass Bottle

Chemical Identifiers

540-84-1
C8H18
114.232
NHTMVDHEPJAVLT-UHFFFAOYSA-N
10907
CHEBI:62805
2,2,4-trimethylpentane
CC(C)CC(C)(C)C

High quality chemicals for laboratory and specialized industrial use

Specific Gravity 1L = 0.69kg
Flash Point −12.00°C
Quantity 4L
Packaging Narrow Mouth Amber Glass Bottle
Formula Weight 114.23
Chemical Name or Material 2,2,4-Trimethylpentane
Grade Baker Analyzed™ ACS Reagent