Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,4-Trimethylpentane, HPLC Grade, Spectrum™

Click to view available options
Quantity:
200 L
Packaging:
Drum
CAS 540-84-1
CAS Max % 1
Grade HPLC
Packaging Drum
Quantity 200 L

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.