Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,5,5-Tetrakis(hydroxymethyl)cyclopentanone, 97%, Acros Organics

Click to view available options
Quantity:
1g
3322-70-1
C9H16O5
204.222
MFCD00223933
ZMFZVJRDJVDKQN-UHFFFAOYSA-N
2,2,5,5-tetrakis hydroxymethyl cyclopentanone,2,2,5,5-tetrakis hydroxymethyl cyclopentan-1-one,2,2,5,5-tetra hydroxymethyl cyclopentanone,cyclopentanone,2,2,5,5-tetrakis hydroxymethyl,2,2,5,5-tetrakis hydroxymethyl-cyclopentanone,2,2,5,5-tetrakis hydroxymethyl-1-cyclopentanone,cyclopentanone, 2,2,5,5-tetrakis hydroxymethyl
234962
2,2,5,5-tetrakis(hydroxymethyl)cyclopentan-1-one
C1CC(C(=O)C1(CO)CO)(CO)CO
3322-70-1
C9H16O5
204.222
MFCD00223933
ZMFZVJRDJVDKQN-UHFFFAOYSA-N
2,2,5,5-tetrakis hydroxymethyl cyclopentanone,2,2,5,5-tetrakis hydroxymethyl cyclopentan-1-one,2,2,5,5-tetra hydroxymethyl cyclopentanone,cyclopentanone,2,2,5,5-tetrakis hydroxymethyl,2,2,5,5-tetrakis hydroxymethyl-cyclopentanone,2,2,5,5-tetrakis hydroxymethyl-1-cyclopentanone,cyclopentanone, 2,2,5,5-tetrakis hydroxymethyl
234962
2,2,5,5-tetrakis(hydroxymethyl)cyclopentan-1-one
C1CC(C(=O)C1(CO)CO)(CO)CO
Quantity 1g
Color White
Melting Point 138°C to 142°C
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 204.22
Physical Form Crystalline Powder
Percent Purity ≥96.0%
Chemical Name or Material 2,2,5,5-Tetrakis(hydroxymethyl)cyclopentanone
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.