Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2':6',2''-Terpyridine, 96%, ACROS Organics™

Click to view available options
Quantity:
5g
1148-79-4
C15H11N3
233.28
MFCD00006213
DRGAZIDRYFYHIJ-UHFFFAOYSA-N
2,2':6',2-terpyridine,tripyridyl,tripyridine,2,6-bis 2-pyridyl pyridine,terpy,terpyridine,2,2',2-terpyridine,2,2',2-terpyridyl,2,2',2-tripyridyl,2,2',2-tripyridine
70848
CHEBI:245199
2,6-dipyridin-2-ylpyridine
C1=CC=NC(=C1)C2=NC(=CC=C2)C3=CC=CC=N3
1148-79-4
C15H11N3
233.28
MFCD00006213
DRGAZIDRYFYHIJ-UHFFFAOYSA-N
2,2':6',2-terpyridine,tripyridyl,tripyridine,2,6-bis 2-pyridyl pyridine,terpy,terpyridine,2,2',2-terpyridine,2,2',2-terpyridyl,2,2',2-tripyridyl,2,2',2-tripyridine
70848
CHEBI:245199
2,6-dipyridin-2-ylpyridine
C1=CC=NC(=C1)C2=NC(=CC=C2)C3=CC=CC=N3
Concentration or Composition (by Analyte or Components) Weight% 100
Odor Odorless
Melting Point 89.0°C to 91.0°C
Color Brown to Cream
Quantity 5g
Boiling Point 370.0°C
Assay Percent Range 95% min. (GC)
UN Number 2811
Formula Weight 233.28
Physical Form Solid
Percent Purity 96%
Chemical Name or Material 2,2′:6′,2′′-Terpyridine
Show More Show Less

GHS H Statement
Fatal if swallowed.
Fatal in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF exposed or concerned:

GHS Signal Word: Danger

DOTInformation : Hazard Class 6.1

EINECSNumber : 214-559-5

Recommended Storage : Keep containers tightly closed in a dry, cool and well-ventilated place