Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,6,6-Tetramethyl-4-piperidinol 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
250g
Packaging:
Glass bottle
2403-88-5
C9H19NO
157.26
MFCD00005983
VDVUCLWJZJHFAV-UHFFFAOYSA-N
2,2,6,6-tetramethyl-4-piperidinol,lastar a,4-piperidinol, 2,2,6,6-tetramethyl,4-hydroxy-2,2,6,6-tetramethylpiperidine,2,2,6,6-tetramethyl-4-hydroxypiperidine,unii-is23q8egz6,is23q8egz6,2,2,6,6-tetramethyl-piperidin-4-ol,pubchem23863
75471
2,2,6,6-tetramethylpiperidin-4-ol
CC1(CC(CC(N1)(C)C)O)C
2403-88-5
C9H19NO
157.26
MFCD00005983
VDVUCLWJZJHFAV-UHFFFAOYSA-N
2,2,6,6-tetramethyl-4-piperidinol,lastar a,4-piperidinol, 2,2,6,6-tetramethyl,4-hydroxy-2,2,6,6-tetramethylpiperidine,2,2,6,6-tetramethyl-4-hydroxypiperidine,unii-is23q8egz6,is23q8egz6,2,2,6,6-tetramethyl-piperidin-4-ol,pubchem23863
75471
2,2,6,6-tetramethylpiperidin-4-ol
CC1(CC(CC(N1)(C)C)O)C
Boiling Point 212°C to 215°C
Color Beige or White
Infrared Spectrum Authentic
Quantity 25g
Melting Point 129°C to 133°C
Packaging Glass bottle
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Beilstein 21, 12
Solubility Information Solubility in water: 130g/l (23 c)
Formula Weight 157.26
Physical Form Crystals, Crystalline Powder and/or Chunks
Percent Purity ≥98.5%
Chemical Name or Material 2,2,6,6-Tetramethyl-4-piperidinol, 99%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 219-291-2

TSCA : TSCA