Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,6,6-Tetramethyl-4-piperidone Hydrochloride 95.0+%, TCI America™

Click to view available options
Quantity:
25g
33973-59-0
C9H18ClNO
191.699
MFCD00012768
ZXNWYMNKYXUZGM-UHFFFAOYSA-N
2,2,6,6-Tetramethyl-4-azacyclohexanone Hydrochloride, Triacetonamine Hydrochloride
94452
2,2,6,6-tetramethylpiperidin-4-one;hydrochloride
CC1(CC(=O)CC(N1)(C)C)C.Cl
33973-59-0
C9H18ClNO
191.699
MFCD00012768
ZXNWYMNKYXUZGM-UHFFFAOYSA-N
2,2,6,6-Tetramethyl-4-azacyclohexanone Hydrochloride, Triacetonamine Hydrochloride
94452
2,2,6,6-tetramethylpiperidin-4-one;hydrochloride
CC1(CC(=O)CC(N1)(C)C)C.Cl
Quantity 25g
Color Undesignated
Formula Weight 191.70
Physical Form Crystal-Powder at 20°C
Percent Purity ≥95.0% (T)
Chemical Name or Material 2,2,6,6-Tetramethyl-4-piperidone Hydrochloride