Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,6,6-Tetramethylpiperidine, 98%, ACROS Organics™

Click to view available options
Quantity:
10g
25g
100g
Packaging:
Glass bottle
768-66-1
C9H19N
141.26
RKMGAJGJIURJSJ-UHFFFAOYSA-N
norpempidine,piperidine, 2,2,6,6-tetramethyl,2,2,6,6-tetramethyl piperidine,unii-44n9s1ycfm,2,2,6,6-tetramethylpeperidine,44n9s1ycfm,2,2,6,6 tetramethyl piperidine,2,2,6,6-tetramethyl-piperidine,pubchem7259,acmc-209p6i
13035
2,2,6,6-tetramethylpiperidine
CC1(CCCC(N1)(C)C)C
768-66-1
C9H19N
141.26
RKMGAJGJIURJSJ-UHFFFAOYSA-N
norpempidine,piperidine, 2,2,6,6-tetramethyl,2,2,6,6-tetramethyl piperidine,unii-44n9s1ycfm,2,2,6,6-tetramethylpeperidine,44n9s1ycfm,2,2,6,6 tetramethyl piperidine,2,2,6,6-tetramethyl-piperidine,pubchem7259,acmc-209p6i
13035
2,2,6,6-tetramethylpiperidine
CC1(CCCC(N1)(C)C)C
Color Colorless to Yellow
Quantity 25g
Packaging Glass bottle
Infrared Spectrum Authentic
Boiling Point 152.0°C
Specific Gravity 0.83
Refractive Index 1.4430 to 1.4460
Flash Point 24°C
Density 0.8300g/mL
Assay Percent Range 98%
Beilstein 20,129
Solubility Information Solubility in water: miscible. Other solubilities: soluble in toluene, dimethylformamide
Formula Weight 141.26
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 2,2,6,6-Tetramethylpiperidine
Show More Show Less