Learn More
Alfa Aesar™ 2-(2-Amino-2-propyl)aniline, 95%
Manufacturer:  Alfa Aesar™ H35906MD
Quantity 250mg
 View more versions of this product
Catalog No. AAH35906MD
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
229326-17-4
150.22
UN2810
2-(2-Amino-2-propyl)aniline
C9H14N2
MFCD08276909
2-Amino-alpha,alpha-dimethylbenzylamine; 2-(2-Aminophenyl)-2-propylamine
Specifications
229326-17-4
C9H14N2
MFCD08276909
2-Amino-alpha,alpha-dimethylbenzylamine; 2-(2-Aminophenyl)-2-propylamine
95%
250mg
150.22
UN2810
2-(2-Amino-2-propyl)aniline
Safety and Handling
SDS & Safety Documents
GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.