Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-(2-Amino-5-nitrophenyl)-4-phenylthiazole, 97%

Click to view available options
Quantity:
250mg
1g
41450-34-4
C15H11N3O2S
297.332
FSEYQHXDVSVHJO-UHFFFAOYSA-N
2-2-amino-5-nitrophenyl-4-phenylthiazole,4-nitro-2-4-phenylthiazol-2-yl aniline,4-nitro-2-4-phenyl-1,3-thiazol-2-yl aniline
86277504
4-nitro-2-(4-phenyl-1,3-thiazol-2-yl)aniline
C1=CC=C(C=C1)C2=CSC(=N2)C3=C(C=CC(=C3)[N+](=O)[O-])N
41450-34-4
C15H11N3O2S
297.332
FSEYQHXDVSVHJO-UHFFFAOYSA-N
2-2-amino-5-nitrophenyl-4-phenylthiazole,4-nitro-2-4-phenylthiazol-2-yl aniline,4-nitro-2-4-phenyl-1,3-thiazol-2-yl aniline
86277504
4-nitro-2-(4-phenyl-1,3-thiazol-2-yl)aniline
C1=CC=C(C=C1)C2=CSC(=N2)C3=C(C=CC(=C3)[N+](=O)[O-])N
Quantity 1g
Melting Point 188°C to 189°C
Color Yellow
Formula Weight 297.33
Physical Form Crystalline Solid
Percent Purity 97%
Chemical Name or Material 2-(2-Amino-5-nitrophenyl)-4-phenylthiazole