Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-(2-Amino-5-nitrophenyl)-5-methyl-4-phenylthiazole, 97%

Click to view available options
Quantity:
250mg
1g
1965305-02-5
C16H13N3O2S
311.359
UCBKPBJMEXATSY-UHFFFAOYSA-N
2-2-amino-5-nitrophenyl-5-methyl-4-phenylthiazole,2-5-methyl-4-phenyl-1,3-thiazol-2-yl-4-nitroaniline
86277505
2-(5-methyl-4-phenyl-1,3-thiazol-2-yl)-4-nitroaniline
CC1=C(N=C(S1)C2=C(C=CC(=C2)[N+](=O)[O-])N)C3=CC=CC=C3
1965305-02-5
C16H13N3O2S
311.359
UCBKPBJMEXATSY-UHFFFAOYSA-N
2-2-amino-5-nitrophenyl-5-methyl-4-phenylthiazole,2-5-methyl-4-phenyl-1,3-thiazol-2-yl-4-nitroaniline
86277505
2-(5-methyl-4-phenyl-1,3-thiazol-2-yl)-4-nitroaniline
CC1=C(N=C(S1)C2=C(C=CC(=C2)[N+](=O)[O-])N)C3=CC=CC=C3
Quantity 250mg
Color Yellow
Formula Weight 311.36
Physical Form Crystalline Solid
Percent Purity 97%
Chemical Name or Material 2-(2-Amino-5-nitrophenyl)-5-methyl-4-phenylthiazole