Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2'-Biphenol, 99%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
Packaging:
Plastic bottle
1806-29-7
C12H10O2
186.21
MFCD00002210
IMHDGJOMLMDPJN-UHFFFAOYSA-N
2,2'-biphenol,1,1'-biphenyl-2,2'-diol,2,2'-dihydroxybiphenyl,biphenyl-2,2'-diol,o,o'-diphenol,o,o'-biphenol,o-dihydroxydiphenyl,o,o'-dihydroxybiphenyl,2,2'-biphenyldiol,2,2'-dihydroxydiphenyl
15731
CHEBI:28970
2-(2-hydroxyphenyl)phenol
C1=CC=C(C(=C1)C2=CC=CC=C2O)O
1806-29-7
C12H10O2
186.21
MFCD00002210
IMHDGJOMLMDPJN-UHFFFAOYSA-N
2,2'-biphenol,1,1'-biphenyl-2,2'-diol,2,2'-dihydroxybiphenyl,biphenyl-2,2'-diol,o,o'-diphenol,o,o'-biphenol,o-dihydroxydiphenyl,o,o'-dihydroxybiphenyl,2,2'-biphenyldiol,2,2'-dihydroxydiphenyl
15731
CHEBI:28970
2-(2-hydroxyphenyl)phenol
C1=CC=C(C(=C1)C2=CC=CC=C2O)O
Density 0.8300g/mL
Boiling Point 315°C
Flash Point 154°C
Specific Gravity 0.83
Color Beige to Beige-Gray
Quantity 100g
Melting Point 108°C to 111°C
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Packaging Plastic bottle
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula HOC6H4C6H4OH
Beilstein 06, 989
Solubility Information Solubility in water: insoluble
Formula Weight 186.21
Physical Form Powder
Percent Purity ≥98.5%
Chemical Name or Material 2,2′-Biphenol, 99%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 217-303-0

RTECSNumber : DV4200000

TSCA : TSCA