Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,2'-Bipyridine-4,4'-dicarboxylic acid, 98%

Click to view available options
Quantity:
1g
5g
25g
6813-38-3
C12H8N2O4
244.206
MFCD00015430
FXPLCAKVOYHAJA-UHFFFAOYSA-N
2,2'-bipyridine-4,4'-dicarboxylic acid,2,2'-bipyridyl-4,4'-dicarboxylic acid,4,4'-dicarboxy-2,2'-bipyridine,2,2'-biisonicotinic acid,2,2'-dipyridyl-4,4'-dicarboxylic acid,2,2'-bipyridine-4,4'-dicarboxylicacid,2-4-carboxypyridin-2-yl pyridine-4-carboxylic acid,4,4-dicarboxy-2,2-bipyridine,2-4-carboxy-2-pyridyl pyridine-4-carboxylic acid
688094
2-(4-carboxypyridin-2-yl)pyridine-4-carboxylic acid
C1=CN=C(C=C1C(=O)O)C2=NC=CC(=C2)C(=O)O
6813-38-3
C12H8N2O4
244.206
MFCD00015430
FXPLCAKVOYHAJA-UHFFFAOYSA-N
2,2'-bipyridine-4,4'-dicarboxylic acid,2,2'-bipyridyl-4,4'-dicarboxylic acid,4,4'-dicarboxy-2,2'-bipyridine,2,2'-biisonicotinic acid,2,2'-dipyridyl-4,4'-dicarboxylic acid,2,2'-bipyridine-4,4'-dicarboxylicacid,2-4-carboxypyridin-2-yl pyridine-4-carboxylic acid,4,4-dicarboxy-2,2-bipyridine,2-4-carboxy-2-pyridyl pyridine-4-carboxylic acid
688094
2-(4-carboxypyridin-2-yl)pyridine-4-carboxylic acid
C1=CN=C(C=C1C(=O)O)C2=NC=CC(=C2)C(=O)O
Applications
2,2′-Bipyridine-4,4′-dicarboxylic acid is used as an organic chemical synthesis intermediate.

Solubility
Insoluble in water. Soluble in common organic solvents.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Melting Point >310°C
Quantity 5g
Formula Weight 244.21
Percent Purity 98%
Chemical Name or Material 2,2'-Bipyridine-4,4'-dicarboxylic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning