2,2'-Bipyridine, BAKER ANALYZED™ Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

Manufacturer: J.T.Baker C32303

Packaging Wide Mouth Amber Glass Bottle
Quantity 25g

 View more versions of this product

Catalog No. 02-002-065

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 366-18-7
    Molecular Formula C10H8N2
    Formula Weight 156.19
    Chemical Name or Material 2,2'-Bipyridine
    Grade BAKER ANALYZED™ Reagent

    Specifications

    Formula Weight 156.19
    Molecular Formula C10H8N2
    Packaging Wide Mouth Amber Glass Bottle
    CAS 366-18-7
    Quantity 25g
    Chemical Name or Material 2,2'-Bipyridine
    Grade BAKER ANALYZED™ Reagent

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit