Alfa Aesar™ 2,2-Bis[4-(4-aminophenoxy)phenyl]propane, 98%

Manufacturer: Alfa Aesar™ H5505722

Quantity 100g

 View more versions of this product

Catalog No. AAH5505722

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13080-86-9
    Molecular Formula C27H26N2O2
    Formula Weight 410.52
    MDL Number MFCD00039152
    Synonym 4,4'-(4,4'-Isopropylidenediphenyl-1,1'-diyldioxy)dianiline; Bisphenol A bis(4-aminophenyl) ether
    Chemical Name or Material 2,2-Bis[4-(4-aminophenoxy)phenyl]propane

    Specifications

    Formula Weight 410.52
    MDL Number MFCD00039152
    Melting Point 127° to 130°C
    Quantity 100g
    RTECS Number CY1062500
    TSCA TSCA
    Molecular Formula C27H26N2O2
    CAS 13080-86-9
    Synonym 4,4'-(4,4'-Isopropylidenediphenyl-1,1'-diyldioxy)dianiline; Bisphenol A bis(4-aminophenyl) ether
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material 2,2-Bis[4-(4-aminophenoxy)phenyl]propane
    Assay 98%

    Description

    Applications
    It is applied in pharmaceutical industry.

    Solubility
    Very slightly soluble in water. Soluble in acetone, Dimethyl sulfoxide(DMSO).

    Notes
    Store at room temperature with proper ventilation. Keep away from oxidizing agents.

    GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit