Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,2-Bis(4-methoxyphenyl)ethylamine, 98%

Click to view available options
Quantity:
1g
5g
85336-83-0
C16H19NO2
257.333
MFCD04039769
VVTAUYUSSABNDB-UHFFFAOYSA-N
2,2-bis 4-methoxyphenyl ethylamine,2,2-bis 4-methoxyphenyl ethanamine,2,2-bis 4-methoxylphenyl ethylamine,benzeneethanamine,4-methoxy-b-4-methoxyphenyl
7016418
2,2-bis(4-methoxyphenyl)ethanamine
COC1=CC=C(C=C1)C(CN)C2=CC=C(C=C2)OC
85336-83-0
C16H19NO2
257.333
MFCD04039769
VVTAUYUSSABNDB-UHFFFAOYSA-N
2,2-bis 4-methoxyphenyl ethylamine,2,2-bis 4-methoxyphenyl ethanamine,2,2-bis 4-methoxylphenyl ethylamine,benzeneethanamine,4-methoxy-b-4-methoxyphenyl
7016418
2,2-bis(4-methoxyphenyl)ethanamine
COC1=CC=C(C=C1)C(CN)C2=CC=C(C=C2)OC
Melting Point 45°C to 50°C
Quantity 5g
Formula Weight 257.34
Percent Purity 98%
Chemical Name or Material 2,2-Bis(4-methoxyphenyl)ethylamine

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning