Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2-Bis(hydroxymethyl)propionic acid, 98%, Acros Organics™

Click to view available options
Quantity:
500g
1kg
5kg
Packaging:
Plastic bottle
Plastic drum
4767-03-7
C5H10O4
134.13
PTBDIHRZYDMNKB-UHFFFAOYSA-N
2,2-bis hydroxymethyl propionic acid,dimethylolpropionic acid,3-hydroxy-2-hydroxymethyl-2-methylpropanoic acid,2,2-bis hydroxymethyl propanoic acid,propanoic acid, 3-hydroxy-2-hydroxymethyl-2-methyl,2,2-dimethylolpropionic acid,dimethylol propionic acid,2,2-bis hydroxymethyl propanoicacid,unii-4nhi8v17mn,4nhi8v17mn
78501
3-hydroxy-2-(hydroxymethyl)-2-methylpropanoic acid
CC(CO)(CO)C(=O)O
4767-03-7
C5H10O4
134.13
PTBDIHRZYDMNKB-UHFFFAOYSA-N
2,2-bis hydroxymethyl propionic acid,dimethylolpropionic acid,3-hydroxy-2-hydroxymethyl-2-methylpropanoic acid,2,2-bis hydroxymethyl propanoic acid,propanoic acid, 3-hydroxy-2-hydroxymethyl-2-methyl,2,2-dimethylolpropionic acid,dimethylol propionic acid,2,2-bis hydroxymethyl propanoicacid,unii-4nhi8v17mn,4nhi8v17mn
78501
3-hydroxy-2-(hydroxymethyl)-2-methylpropanoic acid
CC(CO)(CO)C(=O)O
Color White
Water 0.5% Max. (K.F.)
Quantity 1kg
Melting Point 185.0°C to 190.0°C
Infrared Spectrum Authentic
Packaging Plastic bottle
Flash Point >150°C
Assay Percent Range 98%
Linear Formula CH3C(CH2OH)2CO2H
Beilstein 03,401
Merck Index 15,3277
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol,slightly soluble in acetone,insoluble in benzene
Formula Weight 134.13
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 2, 2-Bis(hydroxymethyl)propionic acid
Show More Show Less

GHS P Statement
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning