Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(2-Bromoethoxy)tetrahydro-2H-pyran, 96%, stabilized, Thermo Scientific Chemicals

Catalog Number p-3771021
Click to view available options
Quantity:
25 mL
Packaging:
Glass bottle
17739-45-6
C7H13BrO2
209.08
MFCD01321310
GCUOLJOTJRUDIZ-UHFFFAOYSA-N
2-2-bromoethoxy tetrahydro-2h-pyran, 2-2-bromoethoxy oxane, 2h-pyran, 2-2-bromoethoxy tetrahydro, 2-2-bromo-ethoxy-tetrahydro-pyran, 2-2-bromoethoxy tetrahydropyran, 2 2-bromoethoxy tetrahydro-2h-pyran, 2-2-bromoethoxy-tetrahydro-2h-pyran, 2h-pyran,3-2-bromoethoxy tetrahydro, 2-2-bromoethyl oxy tetrahaydro-2h-pyran
86621
2-(2-bromoethoxy)oxane
C1CCOC(C1)OCCBr
17739-45-6
C7H13BrO2
209.08
MFCD01321310
GCUOLJOTJRUDIZ-UHFFFAOYSA-N
2-2-bromoethoxy tetrahydro-2h-pyran, 2-2-bromoethoxy oxane, 2h-pyran, 2-2-bromoethoxy tetrahydro, 2-2-bromo-ethoxy-tetrahydro-pyran, 2-2-bromoethoxy tetrahydropyran, 2 2-bromoethoxy tetrahydro-2h-pyran, 2-2-bromoethoxy-tetrahydro-2h-pyran, 2h-pyran,3-2-bromoethoxy tetrahydro, 2-2-bromoethyl oxy tetrahaydro-2h-pyran
86621
2-(2-bromoethoxy)oxane
C1CCOC(C1)OCCBr

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 95.0
CAS Max % 100.0
Density 1.3840g/mL
Boiling Point 62°C to 64°C (0.4 mmHg)
Flash Point 87°C
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Quantity 25 mL
Specific Gravity 1.384
Solubility Information Solubility in water: insoluble
Formula Weight 209.08
Percent Purity 96%
Chemical Name or Material 2-(2-Bromoethoxy)tetrahydro-2H-pyran
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit