Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(2-Bromoethyl)-1,3-dioxolane, 96%, ACROS Organics™

Click to view available options
Quantity:
10g
25g
50g
Packaging:
Glass bottle
18742-02-4
C5H9BrO2
181.03
MFCD00003216
GGZQLTVZPOGLCC-UHFFFAOYSA-N
2-2-bromoethyl-1,3-dioxolane,1,3-dioxolane, 2-2-bromoethyl,bromoethyl-1,3-dioxolane,acmc-1bol0,2-2-bromoethyl-dioxolane,ksc496m8b,2-bromoethyl-1,3-dioxolane,2 2-bromoethyl-1,3-dioxolane,2-2-bromoethyl-1,3-dioxolan
87776
2-(2-bromoethyl)-1,3-dioxolane
C1COC(O1)CCBr
18742-02-4
C5H9BrO2
181.03
MFCD00003216
GGZQLTVZPOGLCC-UHFFFAOYSA-N
2-2-bromoethyl-1,3-dioxolane,1,3-dioxolane, 2-2-bromoethyl,bromoethyl-1,3-dioxolane,acmc-1bol0,2-2-bromoethyl-dioxolane,ksc496m8b,2-bromoethyl-1,3-dioxolane,2 2-bromoethyl-1,3-dioxolane,2-2-bromoethyl-1,3-dioxolan
87776
2-(2-bromoethyl)-1,3-dioxolane
C1COC(O1)CCBr
Boiling Point 68°C to 70°C (8.0mmHg)
Specific Gravity 1.54
Flash Point 65°C
Refractive Index 1.4783 to 1.4803
Packaging Glass bottle
Quantity 25g
Density 1.5400g/mL
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 95% min. (GC)
Fieser 07,37; 11,78
Solubility Information Solubility in water: immiscible
Formula Weight 181.03
Percent Purity 96%
Chemical Name or Material 2-(2-Bromoethyl)-1, 3-dioxolane
Show More Show Less

GHS H Statement
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attent

GHS Signal Word: Warning

EINECSNumber : 242-551-1