Alfa Aesar™ 2-(2-Chloro-4-methoxyphenyl)-3-oxobutyronitrile, 95%

Manufacturer: Alfa Aesar™ H26022MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH26022MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 441060-95-3
    Molecular Formula C11H10ClNO2
    Formula Weight 223.66
    MDL Number MFCD08445711
    UN Number UN3439
    Synonym alpha-Acetyl-2-chloro-4-methoxyphenylacetonitrile
    Chemical Name or Material 2-(2-Chloro-4-methoxyphenyl)-3-oxobutyronitrile

    Specifications

    Formula Weight 223.66
    Molecular Formula C11H10ClNO2
    MDL Number MFCD08445711
    UN Number UN3439
    Synonym alpha-Acetyl-2-chloro-4-methoxyphenylacetonitrile
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H331-H302-H312-H315-H319-H335
    Toxic if inhaled.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Melting Point 78° to 80°C
    Quantity 250mg
    CAS 441060-95-3
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
    Chemical Name or Material 2-(2-Chloro-4-methoxyphenyl)-3-oxobutyronitrile
    Assay 95%

    GHS H Statement
    H331-H302-H312-H315-H319-H335
    Toxic if inhaled.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit