Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-(2-Chlorophenoxy)acetamide, 97%

Click to view available options
Quantity:
1g
5g
25g
35368-68-4
C8H8ClNO2
185.607
MFCD00173819
VFUNLXYSSCTXBG-UHFFFAOYSA-N
2-2-chlorophenoxy acetamide,acmc-1ago0,2-chlorophenoxy acetamide,aronis24002
97329
2-(2-chlorophenoxy)acetamide
C1=CC=C(C(=C1)OCC(=O)N)Cl
35368-68-4
C8H8ClNO2
185.607
MFCD00173819
VFUNLXYSSCTXBG-UHFFFAOYSA-N
2-2-chlorophenoxy acetamide,acmc-1ago0,2-chlorophenoxy acetamide,aronis24002
97329
2-(2-chlorophenoxy)acetamide
C1=CC=C(C(=C1)OCC(=O)N)Cl
Quantity 5g
Formula Weight 185.61
Percent Purity 97%
Chemical Name or Material 2-(2-Chlorophenoxy)acetamide

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning